  • Search Results
  • Phytochemical profile, antioxidative and anti-inflammatory potentials of Gynura bicolor DC.
Phytochemical profile, antioxidative and anti-inflammatory potentials of Gynura bicolor DC.
Journal of the science of food and agriculture (2014-09-10)
Che-yi Chao, Wen-hu Liu, Jia-jiuan Wu, Mei-chin Yin
ABSTRACT
The phytochemical composition of aqueous and ethanol extracts from Gynura bicolor DC., a vegetable, was determined. Human umbilical vein endothelial (HUVE) cells were used to examine the antioxidative and anti-inflammatory potentials of these extracts at 1, 2 or 4% (v/v) against high-glucose-induced injury. Both aqueous and ethanol extracts contained phenolic acids, flavonoids, carotenoids and anthocyanins in the ranges 1428-1569, 1934-2175, 921-1007 and 2135-2407 mg per 100 g dry weight respectively. Both extracts were rich in quercetin, lutein, malvidin and pelargonidin. Addition of these extracts at test doses decreased reactive oxygen species formation, preserved glutathione content and retained glutathione peroxide and catalase activities in high-glucose-treated HUVE cells (P < 0.05). Treatments with these extracts at 2 and 4% lowered interleukin-6, tumor necrosis factor-alpha and prostaglandin E2 production and reduced cyclooxygenase-2 activity (P < 0.05). These findings suggest that this vegetable could be considered as a functional food and might provide antioxidative and anti-inflammatory protection.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
MTT Formazan, powder
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
MTT Formazan, MTT reduction end product
Sigma-Aldrich
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C