  • Search Results
  • Bone metastasis imaging with SPECT/CT/MRI: a preclinical toolbox for therapy studies.
Bone metastasis imaging with SPECT/CT/MRI: a preclinical toolbox for therapy studies.
Bone (2015-02-15)
Pedro Gomes Sanches, Steffie Peters, Raffaella Rossin, Eric L Kaijzel, Ivo Que, Clemens W G M Löwik, Holger Grüll
ABSTRACT
Bone is one of the most common metastatic target sites in breast cancer, with more than 200 thousand new cases of invasive cancer diagnosed in the US alone in 2011. We set out to establish a multimodality imaging platform for bone metastases in small animals as a tool to non-invasively quantify metastasis growth, imaging the ensuing bone lesions and possibly the response to treatment. To this end, a mouse model of osteolytic metastatic bone tumors was characterized with SPECT/CT and MRI over time. A cell line capable of forming bone metastases, MDA-MB-231, was genetically modified to stably express the reporter gene herpes simplex virus-1 thymidine kinase (hsv-1 tk). The intracellular accumulation of the radiolabeled tracer [(123)I]FIAU promoted by HSV-1 TK specifically pinpoints the location of tumor cells which can be imaged in vivo by SPECT. First, a study using tumors implanted subcutaneously was performed. The SPECT/MRI overlays and the ex vivo γ-counting showed a linear correlation in terms of %ID/cm(3) (R(2)=0.93) and %ID/g (R(2)=0.77), respectively. Then, bone metastasis growth was imaged weekly by SPECT/CT and T2-weighted MRI over a maximum of 40 days post-intracardiac injection of tumor cells. The first activity spots detectable with SPECT, around day 20 post-cell injection, were smaller than 2mm(3) and not yet visible by MRI and increased in volume and in %ID over the weeks. Osteolytic bone lesions were visible by CT (in vivo) and μCT (ex vivo). The SPECT/MRI overlays also showed a linear correlation in terms of %ID/cm(3) (R(2)=0.86). In conclusion, a new multimodality imaging platform has been established that non-invasively combines images of active tumor areas (SPECT), tumor volume (MRI) and the corresponding bone lesions (CT and μCT). To our knowledge this is the first report where the combination of soft tissue information from MRI, bone lesions by CT, and reporter gene imaging by SPECT is used to non-invasively follow metastatic bone lesions.
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Uracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Ganciclovir, ≥99% (HPLC), powder
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Lutetium(III) chloride, anhydrous, powder, 99.99% trace metals basis