  • Search Results
  • One-step solvent extraction followed by liquid chromatography-atmospheric pressure photoionization tandem mass spectrometry for the determination of polycyclic aromatic hydrocarbons in edible oils.
One-step solvent extraction followed by liquid chromatography-atmospheric pressure photoionization tandem mass spectrometry for the determination of polycyclic aromatic hydrocarbons in edible oils.
Analytical and bioanalytical chemistry (2015-03-03)
Long-Kai Shi, Yu-Lan Liu, Hua-Min Liu, Ming-Ming Zhang
ABSTRACT
A method for rapid analysis of 16 polycyclic aromatic hydrocarbons (PAHs) in edible oils has been developed on the basis of a simplified solvent extraction and liquid chromatography-atmospheric pressure photoionization tandem mass spectrometry performed in multiple reaction monitoring mode. The briefness of the experimental procedure, the use of milliliters of acetonitrile for extraction without any further cleanup process, the short analysis time, and the excellent sensitivity and selectivity demonstrated the advantages of this practical and environmentally friendly method. All the analytes exhibited satisfactory recoveries at three spiking levels (the recoveries ranged from 77.8 to 106.4%), and the relative standard deviations were lower than 10%. The limits of quantitation of this method for the 16 PAHs were in the range of 0.02-0.43 μg/kg. The validated method was successfully applied for the determination of PAHs in coconut oil reference material (BCR-458) and real edible oil samples. The results suggested that a large-scale investigation of the concentration of PAHs in vegetable oils in China is required.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorobenzene, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phenanthrene, 98%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acenaphthene, 99%
Supelco
Toluene, HPLC grade, 99.8%
Supelco
Toluene, analytical standard
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Acenaphthene, analytical standard
Supelco
Phenanthrene, analytical standard, for environmental analysis
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard