MicroRNA-101 inhibits rat cardiac hypertrophy by targeting Rab1a.
Journal of cardiovascular pharmacology (2015-04-09)
Longxiao Wei, Menghui Yuan, Runsuo Zhou, Qianrong Bai, Wei Zhang, Ming Zhang, Yong Huang, Le Shi
ABSTRACT
Cardiac hypertrophy is a primary pathological change associated with cardiovascular diseases. Dysregulated microRNAs are frequent in cardiovascular diseases and contribute to cardiac hypertrophy by regulating a series of targeted genes. In this study, a rat model of cardiac hypertrophy was created by transverse abdominal aortic constriction, and cardiomyocyte hypertrophy in cultured neonatal rat cardiomyocytes was induced using angiotensin II (AngII) to investigate the role of miR-101 in myocardial hypertrophy. We demonstrated that miR-101 was downregulated in both the transverse abdominal aortic constriction rat model and hypertrophic cardiac myocytes. The overexpression of miR-101 in neonatal rat cardiomyocytes, which was accompanied by a reduced Rab1a level, inhibits 3 cardinal features of cardiomyocyte hypertrophy: fetal gene expression, protein synthesis, and cell enlargement. Conversely, the downregulation of miR-101 reverses these effects. Furthermore, the luciferase reporter system demonstrated that Rab1a is a target gene of miR-101, and the ectopic expression of Rab1a can reverse the cardiomyocyte hypertrophy inhibitory activity of miR-101. Taken together, our findings identify miR-101 as an important regulator in cardiac hypertrophy and implicate the potential application of miR-101 in the therapy of cardiac hypertrophy.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis