  • Search Results
  • Investigation of the functional expression of purine and pyrimidine receptors in porcine isolated pancreatic arteries.
Investigation of the functional expression of purine and pyrimidine receptors in porcine isolated pancreatic arteries.
Purinergic signalling (2013-12-07)
M Alsaqati, S L F Chan, V Ralevic
ABSTRACT
Receptors for purines and pyrimidines are expressed throughout the cardiovascular system. This study investigated their functional expression in porcine isolated pancreatic arteries. Pancreatic arteries (endothelium intact or denuded) were prepared for isometric tension recording and preconstricted with U46619, a thromboxane A(2) mimetic; adenosine-5'-diphosphate (ADP), uridine-5'-triphosphate (UTP) and MRS2768, a selective P2Y(2) agonist, were applied cumulatively, while adenosine-5'-triphosphate (ATP) and αβ-methylene-ATP (αβ-meATP) response curves were generated from single concentrations per tissue segment. Antagonists/enzyme inhibitors were applied prior to U46619 addition. ATP, αβ-meATP, UTP and MRS2768 induced vasoconstriction, with a potency order of αβ-meATP > MRS2768 > ATP ≥ UTP. Contractions to ATP and αβ-meATP were blocked by NF449, a selective P2X(1) receptor antagonist. The contraction induced by ATP, but not UTP, was followed by vasorelaxation. Endothelium removal and DUP 697, a cyclooxygenase-2 inhibitor, had no significant effect on contraction to ATP but attenuated that to UTP, indicating actions at distinct receptors. MRS2578, a selective P2Y(6) receptor antagonist, had no effect on contractions to UTP. ADP induced endothelium-dependent vasorelaxation which was inhibited by MRS2179, a selective P2Y(1) receptor antagonist, or SCH58261, a selective adenosine A(2A) receptor antagonist. The contractions to ATP and αβ-meATP were attributed to actions at P2X(1) receptors on the vascular smooth muscle, whereas it was shown for the first time that UTP induced an endothelium-dependent vasoconstriction which may involve P2Y(2) and/or P2Y(4) receptors. The relaxation induced by ADP is mediated by P2Y(1) and A(2A) adenosine receptors. Porcine pancreatic arteries appear to lack vasorelaxant P2Y(2) and P2Y(4) receptors.
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.5-100.5%
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%