  • Search Results
  • Perivascular adipose tissue inhibits endothelial function of rat aortas via caveolin-1.
Perivascular adipose tissue inhibits endothelial function of rat aortas via caveolin-1.
PloS one (2014-06-14)
Michelle Hui-Hsin Lee, Shiu-Jen Chen, Cheng-Ming Tsao, Chin-Chen Wu
ABSTRACT
Perivascular adipose tissue (PVAT)-derived factors have been proposed to play an important role in the pathogenesis of atherosclerosis. Caveolin-1 (Cav-1), occupying the calcium/calmodulin binding site of endothelial NO synthase (eNOS) and then inhibiting nitric oxide (NO) production, is also involved in the development of atherosclerosis. Thus, we investigated whether PVAT regulated vascular tone via Cav-1 and/or endothelial NO pathways. Isometric tension studies were carried out in isolated thoracic aortas from Wistar rats in the presence and absence of PVAT. Concentration-response curves of phenylephrine, acetylcholine, and sodium nitroprusside were illustrated to examine the vascular reactivity and endothelial function. The protein expressions of eNOS and Cav-1 were also examined in aortic homogenates. Our results demonstrated that PVAT significantly enhanced vasoconstriction and inhibited vasodilatation via endothelium-dependent mechanism. The aortic NO production was diminished after PVAT treatment, whereas protein expression and activity of eNOS were not significantly affected. In addition, Cav-1 protein expression was significantly increased in aortas with PVAT transfer. Furthermore, a caveolae depleter methyl-β-cyclodextrin abolished the effect of PVAT on the enhancement of vasoconstriction, and reversed the impairment of aortic NO production. In conclusion, unknown factor(s) released from PVAT may inhibit endothelial NO production and induce vasocontraction via an increase of Cav-1 protein expression.
Product Description
Supelco
Sodium Nitroprusside, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
TEMPO, purified by sublimation, 99%
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard