  • Search Results
  • PPP1R1B-STARD3 chimeric fusion transcript in human gastric cancer promotes tumorigenesis through activation of PI3K/AKT signaling.
PPP1R1B-STARD3 chimeric fusion transcript in human gastric cancer promotes tumorigenesis through activation of PI3K/AKT signaling.
Oncogene (2013-11-28)
S M Yun, K Yoon, S Lee, E Kim, S-H Kong, J Choe, J M Kang, T-S Han, P Kim, Y Choi, S Jho, H Yoo, J Bhak, H-K Yang, S-J Kim
ABSTRACT
Fusion genes act as potent oncogenes, resulting from chromosomal rearrangements or abnormal transcription in many human cancers. Although multiple gastric cancer genomes have been sequenced, the driving recurrent gene fusions have not been well characterized. Here, we used paired-end transcriptome sequencing to identify novel gene fusions in 18 human gastric cancer cell lines and 18 pairs of primary human gastric cancer tissues and their adjacent normal tissues. Multiple samples revealed expression of PPP1R1B-STARD3 fusion transcript. The presence of PPP1R1B-STARD3 correlated with elevated levels of PPP1R1B mRNA. PPP1R1B-STARD3 fusion transcript was detected in 21.3% of primary human gastric cancers but not in adjacent matched normal gastric tissues. Based on reverse transcription PCR analysis of DNA, unlike other fusions described in gastric cancer, the PPP1R1B-STARD3 appears to be generated by RNA processing without chromosomal rearrangement. Overexpression of PPP1R1B-STARD3 in MKN-28 significantly increased cell proliferation and colony formation. This increased proliferation was mediated by activation of phosphatidylinositol-3-kinase (PI3K)/AKT signaling. Furthermore, expression of PPP1R1B-STARD3 fusion transcript enhanced the tumor growth of MKN-28 cells in athymic nude mice. These findings show that PPP1R1B-STARD3 fusion transcript has a key role in subsets of gastric cancers through the activation of PI3K/AKT signaling.
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-AKT1 (pThr450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6D6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2H1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6G6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-PKB (pSer473) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Rac antibody produced in mouse, clone 23A8, purified immunoglobulin, buffered aqueous solution