  • Search Results
  • Paclitaxel-induced hyperalgesia modulates negative affective component of pain and NR1 receptor expression in the frontal cortex in rats.
Paclitaxel-induced hyperalgesia modulates negative affective component of pain and NR1 receptor expression in the frontal cortex in rats.
Neuroscience research (2014-01-21)
Kazuko Noda, Hisanao Akita, Masanori Ogata, Makoto Saji
ABSTRACT
Paclitaxel, one of the chemotherapeutic agents clinically used to treat several types of cancer, produces side effects such as peripheral neuropathy, sensory abnormalities, and hyperalgesia. Since hyperalgesia remains after cessation of paclitaxel therapy and becomes chronic, we hypothesize that alteration in memory and the cognitive process of pain underlies hyperalgesia. To test this hypothesis, we examined whether drug-induced hyperalgesia alters the affective component of pain and the NMDA-NR1 and mGluR1 receptors as a mediator for signal transmission and memory of pain. Mechanical sensitivity was measured by von Frey filament test after intraperitoneal injection of paclitaxel in rats. Paclitaxel-induced hyperalgesia was confirmed over almost the entire 14-day period of observation after the treatment. The effect of paclitaxel-induced hyperalgesia on the affective component of pain was assessed using pain-induced place aversion. The formalin-induced conditioned place aversion was completely abolished in the paclitaxel-treated rats. Immunoblot analysis of NR1 and mGluR1 protein levels in various brain regions was performed after paclitaxel treatment. Treatment reduced only the NR1 expression within the frontal cortex. These results suggest that the hypofunction of memory processes with the reduced NMDA receptors in the frontal cortex might be involved in the expression of abnormal emotional behaviors accompanied by hyperalgesia.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Supelco
Pefabloc® SC, analytical standard
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C