  • Search Results
  • Interleukin 6 alters localization of hMSH3, leading to DNA mismatch repair defects in colorectal cancer cells.
Interleukin 6 alters localization of hMSH3, leading to DNA mismatch repair defects in colorectal cancer cells.
Gastroenterology (2014-12-03)
Stephanie S Tseng-Rogenski, Yasushi Hamaya, Daniel Y Choi, John M Carethers
ABSTRACT
Elevated microsatellite alterations at selected tetranucleotide repeats (EMAST) is the most common DNA mismatch repair defect in colorectal cancers, observed in approximately 60% of specimens. This acquired genotype correlates with metastasis and poor outcomes for patients, and is associated with intra-epithelial inflammation and heterogeneous nuclear levels of the mismatch repair protein hMSH3. Inflammation and accompanying oxidative stress can cause hMSH3 to change its intracellular location, but little is known about the source of oxidative stress in cancer cells. We investigated whether cytokines mediate this process. We analyzed levels of interleukin 6 (IL6) and its receptor (IL6R) in human colon and lung cancer cell lines by flow cytometry and enzyme-linked immunosorbent assay; proteins were localized by immunofluorescence and immunoblot analyses. IL6 signaling was blocked with antibodies against IL6, soluble glycoprotein 130 Fc fragments, and the signal transducers and activators of transcription 3 inhibitor NSC74859; a constitutively active form of STAT3 was expressed in colon and lung cancer cell lines to replicate IL6R signaling. EMAST was detected by DNA fragment analysis. Immunohistochemistry was used to examine levels of IL6 in 20 colorectal tumor and adjacent nontumor tissues. Incubation of colon and lung cancer cell lines with IL6, but not other cytokines, caused hMSH3, but no other mismatch repair proteins, to move from the nucleus to the cytosol after generation of oxidative stress; inhibition of IL6 signaling prevented this shift. Expression of constitutively active STAT3 also caused hMSH3 to translocate from the nucleus to the cytoplasm in cancer cell lines. Incubation of cells with IL6 led to tetranucleotide frameshifts, the signature for EMAST. EMAST-positive colorectal tumors had significantly higher levels of IL6 than EMAST-negative tumors. IL6 signaling disrupts the nuclear localization of hMSH3 and DNA repair, leading to EMAST in cancer cell lines. Inflammatory cytokines might therefore promote genetic alterations in human cancer cells.
Sigma-Aldrich
Sodium selenite, 99%
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
9-Ethylcarbazole, 97%
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10