  • Search Results
  • Apple polysaccharide reduces NF-Kb mediated colitis-associated colon carcinogenesis.
Apple polysaccharide reduces NF-Kb mediated colitis-associated colon carcinogenesis.
Nutrition and cancer (2014-11-21)
Dian Zhang, Man Mi, Fengliang Jiang, Yang Sun, Yuhua Li, Libin Yang, Lei Fan, Qian Li, Jin Meng, Zhenggang Yue, Li Liu, Qibing Mei
ABSTRACT
Nuclear factor-kappa B (NF-κB) is an important molecule in mediating inflammatory colitis, which can lead to colorectal cancer (CRC). The aim of this study was to evaluate the chemopreventive efficacy of apple polysaccharide extract (AP) in inhibiting NF-κB-mediated inflammation pathways in CRC. We evaluated AP in vitro in HT-29 and SW620 human CRC cells. We also used the azoxymethane and dextran sodium sulphate (AOM/DSS) model to induce colon carcinogenesis in vivo. The chemoprotective effects of AP were assessed using Western blot, immunofluorescence assay, real-time PCR, electrophoretic mobility shift assay, and flow cytometry. AP reduced AOM/DSS-associated toxicities, prevented carcinogenesis, and decreased the expression of TLR4, MD2, MyD88, TRAM, TRIF-related adapter molecule, interferon-β, tumor necrosis factor-α, and interleukin-6. The protective effects of AP may be related to the inhibition of TLR4/MD2-mediated signaling, including MyD88 and TRIF, as well as the inhibition of NF-κB-mediated inflammatory signaling pathways. Therefore, AP could be used in combination therapy for the prevention of colitis-associated colon cancer.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Azoxymethane, 13.4 M, ≥98%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%