  • Search Results
  • Effect of preoperative simvastatin treatment on transplantation of cryopreserved-warmed mouse ovarian tissue quality.
Effect of preoperative simvastatin treatment on transplantation of cryopreserved-warmed mouse ovarian tissue quality.
Theriogenology (2014-12-03)
Jaewang Lee, Jung Ryeol Lee, Hye Won Youm, Chang Suk Suh, Seok Hyun Kim
ABSTRACT
After the ovarian tissue (OT) transplantation, the ischemia-reperfusion injury causes depletion and apoptosis of follicle. Recent reports stated that simvastatin reduces ischemic damage. Therefore, we used the mouse whole ovarian vitrification and autotransplantation models to investigate the effects of simvastatin. Five-week-old B6D2F1 mice were randomly divided into four groups. Three groups were given simvastatin orally (5 mg/kg) before ovariectomy, either 2 hours before (2H Tx) or once a day for 3 or 7 days. The control group was given saline 2 hours before ovariectomy. All ovaries were cryopreserved by vitrification, held in liquid nitrogen for 1 week before being warmed, and autotransplanted. The grafts were collected for analysis on 2, 7, or 21 days after transplantation. Ovarian follicle morphology and apoptosis were assessed by hematoxylin and eosin staining and terminal deoxynucleotidyl transferase dUTP nick end labeling assay. Vessel integrity in ovary was evaluated by immunohistochemistry using anti-CD31 antibody. Serum FSH level was measured to estimate the transplanted ovarian reserve. The proportion of morphologically normal (G1) follicles at 7 and 21 days and the percentage of CD31 (+) tissue at 21 days was significantly higher in the 2H Tx group than that in the control group. In addition, the 2H Tx group showed a significantly increased intact primordial follicle ratio at 2 and 21 days after OT transplantation. Administration of simvastatin 2 hours before ovariectomy could improve the quality after transplantation of cryopreserved mouse OT.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Hematoxylin
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material