  • Search Results
  • Hypoxia-induced endothelial dysfunction in apolipoprotein E-deficient mice; effects of infliximab and L-glutathione.
Hypoxia-induced endothelial dysfunction in apolipoprotein E-deficient mice; effects of infliximab and L-glutathione.
Atherosclerosis (2014-08-26)
Izabela Tuleta, Carolina Nunes França, Daniela Wenzel, Bernd Fleischmann, Georg Nickenig, Nikos Werner, Dirk Skowasch
ABSTRACT
Obstructive sleep apnoea (OSA) has been implicated as a risk factor for atherosclerosis. The aim of our study was to examine the effects of chronic intermittent hypoxia in apoE-/- mice serving as model of OSA on endothelial dysfunction and oxidative stress and to evaluate the reversibility of hypoxia-induced changes under anti-inflammatory infliximab and anti-oxidative l-glutathione. ApoE-/- mice were divided into 4 groups (n = 9 each): 1. intermittent hypoxia 8 h/day for 6 weeks, 2. intermittent hypoxia + injections of infliximab, 3. intermittent hypoxia + injections of l-glutathione, 4. normoxia = control. Endothelial function was impaired under hypoxia compared to control. Application of infliximab and l-glutathione improved it to a level of control. The percentage of endothelial microparticles increased under hypoxia compared to other groups. Levels of NADPH oxidase 2-derived reactive oxygen species were approximately 9 times higher in the hypoxia group. The number of sca-1/flk-1+ endothelial progenitor cells was higher in bone marrow and lower in blood under hypoxia vs. other groups. Stromal cell derived factor-1alpha- and matrix metalloproteinase-9-dependent release of these cells from bone marrow was attenuated under hypoxia. The number of DilacLDL+/lectin + early outgrowth progenitor cells and that of colony forming units from these cells were higher under hypoxia. Atherosclerotic plaques in the aorta were more frequent under hypoxia and control in comparison with both drug groups. Intermittent hypoxia contributes to endothelial dysfunction by the local increase in reactive oxygen species and reduction of the peripheral repair capacity. Infliximab and l-glutathione prevent hypoxia-induced vascular and extravascular changes.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)