  • Search Results
  • Effects of multiple-dose rifampicin 450 mg on the pharmacokinetics of fexofenadine enantiomers in Japanese volunteers.
Effects of multiple-dose rifampicin 450 mg on the pharmacokinetics of fexofenadine enantiomers in Japanese volunteers.
Journal of clinical pharmacy and therapeutics (2014-09-30)
Y Akamine, M Miura, N Yasui-Furukori, I Ieiri, T Uno
ABSTRACT
Rifampicin is a potent inducer of P-glycoprotein (P-gp) and inhibitor of organic anion-transporting polypeptides (OATPs), with fexofenadine acting as a substrate for both mechanisms. Simultaneous administration of single- or multiple-dose rifampicin 600 mg significantly increases the concentrations of fexofenadine enantiomers by inhibiting OATP transporters. However, the effects of rifampicin 450 mg are unknown. Here, we evaluated the effects of multiple doses of rifampicin 450 mg on the pharmacokinetics of fexofenadine enantiomers in healthy Japanese volunteers. In this randomized, two-phase, double-blind crossover study, 10 healthy volunteers received rifampicin 450 mg/day or placebo for 7 days. On day 7, fexofenadine 60 mg was co-administered simultaneously. Rifampicin significantly increased the mean area under the plasma concentration-time curve (AUC) of (R)- and (S)-fexofenadine (3.10-fold and 3.48-fold, respectively) and decreased the renal clearance of (R)- and (S)-fexofenadine (0.40-fold and 0.47-fold, respectively), causing marked differences in the mean amounts of these enantiomers excreted into the urine in the rifampicin phase (P < 0.001). These results indicated that multiple doses of rifampicin 450 mg may be sufficient to inhibit the renal influx transporter and OATP-mediated hepatic uptake of both enantiomers. Moreover, these effects may be greater than the P-gp-inductive effects of rifampicin. Therefore, the interactive mechanism of multidose rifampicin may occur through a combination of OATP and P-gp transporters, thereby altering the pharmacokinetics of fexofenadine enantiomers. In this study of rifampicin 450 mg, the interactive magnitude of the mean AUC values of fexofenadine enantiomers was higher than that observed in the previous study of rifampicin 600 mg, and no dose-dependent inhibitory effects of rifampicin were observed. These effects may be clinically significant in patients receiving fexofenadine and rifampicin.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Rifampicin, ≥97% (HPLC), powder
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥97% (HPLC), crystalline
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Rifampicin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid