  • Search Results
  • Desferrioxamine as an appropriate chelator for 90Nb: comparison of its complexation properties for M-Df-Octreotide (M = Nb, Fe, Ga, Zr).
Desferrioxamine as an appropriate chelator for 90Nb: comparison of its complexation properties for M-Df-Octreotide (M = Nb, Fe, Ga, Zr).
Nuclear medicine and biology (2014-08-05)
Valery Radchenko, Stefan Busse, Frank Roesch
ABSTRACT
The niobium-90 radioisotope ((90)Nb) holds considerable promise for use in immuno-PET, due to its decay parameters (t½ = 14.6h, positron yield=53%, Eß(+)(mean) = 0.35 MeV and Eß(+)(max) = 1.5 MeV). In particular, (90)Nb appears well suited to detect in vivo the pharmacokinetics of large targeting vectors (50-150 kDa). In order to be useful for immuno-PET chelators are required to both stabilize the radionuclide in terms of coordination chemistry and to facilitate the covalent attachment to the targeting vector. Different chelators were evaluated for this purpose in terms of radiolabelling efficiency and stability of the radiolabelled Nb(V) complex and in order to determine the most suitable candidate for conjugation to a biologically relevant targeting vector. For the purpose of studying the complexation properties the niobium radioisotope (95)Nb was used as an analogue of (90)Nb, by virtue of its longer half-life (35 days) and lower cost (reactor-based production). Acyclic and cyclic chelators were investigated, with desferroxamine [Df: (N'-{5-[acetyl(hydroxy)amino]pentyl}-N-[5-({4-[(5-aminopentyl) (hydroxy)amino]-4-oxobutanoyl} amino)pentyl]-N-hydroxysuccinamide)] emerging as the best candidate. Greater than 99% radiolabelling was achieved at room temperature over a wide pH range. The (95)Nb-Df complex is sufficiently stable for immuno-PET (<7% degradation over 7 days in vitro). As a proof-of-principle, a Df conjugate featuring a well-established targeting vector, (D)-Phe(1)-octreotide, was evaluated. The fast labelling kinetics of the unconjugated chelator (Df) were retained for Df-succinyl-(D)Phe(1)-octreotide (Df-OC), with>90% labelling after 1h at room temperature over the pH range 5-7. Stability studies, performed in vitro in serum at physiological temperature (37 °C), revealed that 87 ± 2% of the radiolabelled molecule remained intact after 7 days. Competition studies with relevant metal ions (zirconium((IV)), gallium((III)) and iron((III))) have been performed with Df-OC to gain insight to the relative stability [Nb-Df]-OC complex to transmetallation. At equimolar metal ion concentrations the [Nb-Df]-OC complex showed the greatest overall stability. The favourable radiolabelling characteristics of Df-OC and its stability indicate that Df is a potentially very useful chelator for the development of radiopharmaceuticals for (90)Nb-PET.
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)