  • Search Results
  • Effects of honokiol on sepsis-induced acute kidney injury in an experimental model of sepsis in rats.
Effects of honokiol on sepsis-induced acute kidney injury in an experimental model of sepsis in rats.
Inflammation (2014-02-18)
Nan Li, Hua Xie, Longkai Li, Jing Wang, Ming Fang, Ning Yang, Hongli Lin
ABSTRACT
Acute kidney injury (AKI) is a severe complication of sepsis, which largely contributes to the high mortality rate of sepsis. Honokiol, a natural product isolated from Magnolia officinalis (Houpo), has been shown to exhibit anti-inflammatory and antioxidant properties. Here, we investigated the effects of honokiol on sepsis-associated AKI in rats subjected to cecal ligation and puncture (CLP). We found that the administration of honokiol improved the survival of septic rats. Periodic acid-Schiff stain revealed that the morphological changes of kidney tissues in CLP rats were restored after honokiol treatment. Furthermore, honokiol reduced CLP-induced oxidative stress and inflammatory cytokine production. The levels of nitric oxide (NO) and inducible NO synthetase (iNOS) were attenuated by honokiol in septic rats. Finally, honokiol inhibited CLP-induced activation of NF-κB signaling in CLP rats. Our findings suggest that honokiol might be used as a potential therapeutic agent for complications of sepsis, especially for sepsis-induced AKI.
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rabbit anti-Histone H3 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Anti-Histone H3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Nitric Oxide Synthase, Inducible antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Histone H3 (Acetyl-Lys18) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-Nitric Oxide Synthase, Inducible antibody produced in mouse, clone NOS-IN, ascites fluid
Sigma-Aldrich
Anti-Histone H3 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Histone H3 (Acetyl-Lys9) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Histone H3 (Acetyl-Lys27) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Histone H3 (Acetyl-Lys23) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-NOS2 antibody produced in mouse, clone 2G4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants