  • Search Results
  • Coordination complexes as molecular glue for immobilization of antibodies on cyclic olefin copolymer surfaces.
Coordination complexes as molecular glue for immobilization of antibodies on cyclic olefin copolymer surfaces.
Analytical biochemistry (2014-04-12)
Huey Wen Ooi, Shaun J Cooper, Chang-Yi Huang, Dean Jennins, Emma Chung, N Joe Maeji, Andrew K Whittaker
ABSTRACT
A novel metal-based chelating method has been used to provide an order of magnitude increase in immunoassay performance on cyclic olefin copolymer (COC) plastics compared with passive binding. COCs are hydrophobic, and without surface modification they are often unsuitable for applications where protein adhesion is desired. When interacting with the bare plastic, the majority of the bound proteins will be denatured and become nonfunctional. Many of the surface modification techniques reported to date require costly equipment setup or the use of harsh reaction conditions. Here, we have successfully demonstrated the use of a simple and quick metal chelation method to increase the sensitivity, activity, and efficiency of protein binding to COC surfaces. A detailed analysis of the COC surfaces after activation with the metal complexes is presented, and the immunoassay performance was studied using three different antibody pairs.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
SAFC
HEPES
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC