  • Search Results
  • Pathophysiological mechanisms underlying increased anxiety after soman exposure: reduced GABAergic inhibition in the basolateral amygdala.
Pathophysiological mechanisms underlying increased anxiety after soman exposure: reduced GABAergic inhibition in the basolateral amygdala.
Neurotoxicology (2014-08-26)
Eric M Prager, Volodymyr I Pidoplichko, Vassiliki Aroniadou-Anderjaska, James P Apland, Maria F M Braga
ABSTRACT
The recent sarin attack in Syria killed 1429 people, including 426 children, and left countless more to deal with the health consequences of the exposure. Prior to the Syrian chemical assault, nerve agent attacks in Japan left many victims suffering from neuropsychiatric illnesses, particularly anxiety disorders, more than a decade later. Uncovering the neuro-pathophysiological mechanisms underlying the development of anxiety after nerve agent exposure is necessary for successful treatment. Anxiety is associated with hyperexcitability of the basolateral amygdala (BLA). The present study sought to determine the nature of the nerve agent-induced alterations in the BLA, which could explain the development of anxiety. Rats were exposed to soman, at a dose that induced prolonged status epilepticus. Twenty-four hours and 14-days after exposure, neurons from the BLA were recorded using whole-cell patch-clamp techniques. At both the 24h and 14-day post-exposure time-points, the frequency and amplitude of spontaneous inhibitory postsynaptic currents (sIPSCs) in the BLA were reduced, along with reduction in the frequency but not amplitude of miniature IPSCs. In addition, activation of α7-nicotinic acetylcholine receptors, a cholinergic receptor that participates in the regulation of BLA excitability and is involved in anxiety, increased spontaneous excitatory postsynaptic currents (sEPSCs) in both soman-exposed rats and controls, but was less effective in increasing sIPSCs in soman-exposed rats. Despite the loss of both interneurons and principal cells after soman-induced status epilepticus, the frequency of sEPSCs was increased in the soman-exposed rats. Impaired function and cholinergic modulation of GABAergic inhibition in the BLA may underlie anxiety disorders that develop after nerve agent exposure.
Sigma-Aldrich
(Aminomethyl)phosphonic acid, 99%
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
Atropine sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(Aminomethyl)phosphonic acid, analytical standard
USP
Atropine sulfate, United States Pharmacopeia (USP) Reference Standard
USP
Choline chloride, United States Pharmacopeia (USP) Reference Standard
Supelco
(Aminomethyl)phosphonic acid, PESTANAL®, analytical standard
Supelco
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid