  • Search Results
  • Oxidored-nitro domain containing protein 1 (NOR1) expression suppresses slug/vimentin but not snail in nasopharyngeal carcinoma: Inhibition of EMT in vitro and in vivo in mice.
Oxidored-nitro domain containing protein 1 (NOR1) expression suppresses slug/vimentin but not snail in nasopharyngeal carcinoma: Inhibition of EMT in vitro and in vivo in mice.
Cancer letters (2014-03-25)
Wei Wang, Xiaoling Li, Wenling Zhang, Wenjuan Li, Mei Yi, Jianbo Yang, Zhaoyang Zeng, Leah E Colvin Wanshura, James B McCarthy, Songqing Fan, Pan Zheng, Shengnan Chen, Bo Xiang, Guiyuan Li
ABSTRACT
Oxidored-nitro domain containing protein 1 (NOR1) is a putative tumor suppressor gene. In this study, NOR1 expression was detected in NPC tissues and non-cancerous nasopharyngeal epithelium. The data showed that NOR1 protein was decreased in NPC tissues. Lost expression NOR1 protein was associated with poor overall and event-free survival of NPC patients. Overexpression of NOR1 in NPC cells resulted in a significant morphological change and decreased expression of epithelial-to-mesenchymal transition (EMT) mediators (e.g., slug and vimentin), but induced cytokeratin 13 expression. A nude mouse metastasis assay revealed that overexpression of NOR1 decreased NPC tumor cells metastasis capacity in vivo. Knockdown of NOR1 expression in HeLa cells was sufficient to abrogate epithelial traits and to enhance cell migration and invasion. Concomitant inhibition of slug or vimentin alleviated induction of EMT, migration or invasion by NOR1 siRNA in HeLa cells in vitro. In conclusion, the data from the current study suggest, for the first time, that NOR1 plays an important role in NPC in ex vivo, in vitro, and in vivo.
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Cdh1 antibody,Mouse monoclonal, clone DCS-266, purified from hybridoma cell culture
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
MISSION® esiRNA, targeting human VIM
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri