  • Search Results
  • The DNA sensor, cyclic GMP-AMP synthase, is essential for induction of IFN-β during Chlamydia trachomatis infection.
The DNA sensor, cyclic GMP-AMP synthase, is essential for induction of IFN-β during Chlamydia trachomatis infection.
Journal of immunology (Baltimore, Md. : 1950) (2014-07-30)
Yugen Zhang, Laxmi Yeruva, Anthony Marinov, Daniel Prantner, Priscilla B Wyrick, Vladimir Lupashin, Uma M Nagarajan
ABSTRACT
IFN-β has been implicated as an effector of oviduct pathology resulting from genital chlamydial infection in the mouse model. In this study, we investigated the role of cytosolic DNA and engagement of DNA sensors in IFN-β expression during chlamydial infection. We determined that three-prime repair exonuclease-1, a host 3' to 5' exonuclease, reduced IFN-β expression significantly during chlamydial infection using small interfering RNA and gene knockout fibroblasts, implicating cytosolic DNA as a ligand for this response. The DNA sensor cyclic GMP-AMP synthase (cGAS) has been shown to bind cytosolic DNA to generate cyclic GMP-AMP, which binds to the signaling adaptor stimulator of IFN genes (STING) to induce IFN-β expression. We determined that cGAS is required for IFN-β expression during chlamydial infection in multiple cell types. Interestingly, although infected cells deficient for STING or cGAS alone failed to induce IFN-β, coculture of cells depleted for either STING or cGAS rescued IFN-β expression. These data demonstrate that cyclic GMP-AMP produced in infected cGAS(+)STING(-) cells can migrate into adjacent cells via gap junctions to function in trans in cGAS(-)STING(+) cells. Furthermore, we observed cGAS localized in punctate regions on the cytosolic side of the chlamydial inclusion membrane in association with STING, indicating that chlamydial DNA is most likely recognized outside the inclusion as infection progresses. These novel findings provide evidence that cGAS-mediated DNA sensing directs IFN-β expression during Chlamydia trachomatis infection and suggest that effectors from infected cells can directly upregulate IFN-β expression in adjacent uninfected cells during in vivo infection, contributing to pathogenesis.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
SAFC
HEPES
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Human IP-10 / CXCL10 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Mouse CRG-2 / CXCL10 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
MISSION® esiRNA, targeting human NTS
Sigma-Aldrich
MISSION® esiRNA, targeting human NT5E