  • Search Results
  • Interleukin-1 beta enhances human multipotent mesenchymal stromal cell proliferative potential and their ability to maintain hematopoietic precursor cells.
Interleukin-1 beta enhances human multipotent mesenchymal stromal cell proliferative potential and their ability to maintain hematopoietic precursor cells.
Cytokine (2014-12-03)
Alexey E Bigildeev, Ekaterina A Zezina, Irina N Shipounova, Nina J Drize
ABSTRACT
Multipotent mesenchymal stromal cells (MMSCs) have been demonstrated to produce mature stromal cells and maintain hematopoietic progenitor cells (HPC). It was previously demonstrated that interleukin-1 beta (IL-1 beta) stimulates the growth of the stromal microenvironment in vivo. The aim of this study was to investigate the effect of IL-1 beta treatment of human MMSCs on their proliferative potential, gene expression, immunomodulating properties, and their ability to support HPCs in vitro. Human bone marrow-derived MMSCs were cultivated in standard conditions or with IL-1 beta. The cumulative cell production was assessed for five passages. After withdrawal of IL-1 beta, MMSC clonal efficiency was investigated, and the maintenance of HPCs on top of MMSCs layers was estimated using cobblestone area forming cell (CAFC) and long-term culture initiating cell (LTC-IC) assays. The effect of untreated MMSCs or MMSCs pretreated with IL-1 beta on lymphocyte proliferation was studied by CFSE staining. The relative expression level of various genes by MMSCs was analyzed using RT-qPCR. The administration of IL-1 beta elevated MMSCs clonal efficiency and total cell production but did not affect lymphocyte proliferation. MMSCs pretreatment with IL-1 beta enhanced their ability to maintain HPCs, as detected by CAFC assay, and it altered the expression levels of genes participating in HPC regulation by stromal cells, e.g., adhesion molecules (ICAM1) and growth factors (SDF1). This study revealed the ability of IL-1 beta to stimulate MMSCs proliferation and enhance their potential to maintain HPCs. MMSCs are considered a stromal niche component in vitro. The combined in vitro and previous in vivo data suggest that IL-1 beta is a systemic regulator of the stromal microenvironment.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard