  • Search Results
  • Directed mouse embryonic stem cells into leydig-like cells rescue testosterone-deficient male rats in vivo.
Directed mouse embryonic stem cells into leydig-like cells rescue testosterone-deficient male rats in vivo.
Stem cells and development (2014-10-24)
Yan Yang, Zhijian Su, Wenting Xu, Jiao Luo, Rui Liang, Qi Xiang, Qihao Zhang, Ren-shan Ge, Yadong Huang
ABSTRACT
The primary function of Leydig cells is to secrete testosterone, which is critical in the regulation of male reproduction and development. Low levels of testosterone will lead to male hypogonadism. Stem cell-derived Leydig cell transplantation may be a promising alternative therapy for male hypogonadism. Thus far, others have reported that Leydig-like cells can be derived from mesenchymal stem cells, embryonic stem cells (ESCs), and induced pluripotent stem cells. However, the efficiency of the differentiating Leydig cells remains low, and progress toward generating functional adult Leydig cells (ALCs) is limited. Herein, we describe a robust method of directing differentiation of mouse embryonic stem cells (mESCs) into Leydig-like cells in vitro by overexpression of the transcription factor steroidogenic factor-1 (SF-1) and treatment with a combination of 8-Bromoadenosine-3',5'-cyclic monophosphate and forskolin. These differentiated cells express mRNA encoding the steroidogenic enzymes and produce progesterone and testosterone. Importantly, when transplanted into male rats that had their original Leydig cells selectively eliminated by ethylene dimethanesulfonate, these in vitro-derived Leydig-like cells further developed into functional ALCs that rescued serum testosterone levels. These data provide evidence that mESCs can be induced to differentiate into Leydig-like cells in vitro, which can develop in the in vivo microenvironment.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethyl sulfoxide, analytical standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water