  • Search Results
  • In vitro and in vivo effects of xanthorrhizol on human breast cancer MCF-7 cells treated with tamoxifen.
In vitro and in vivo effects of xanthorrhizol on human breast cancer MCF-7 cells treated with tamoxifen.
Journal of pharmacological sciences (2014-08-22)
Nattanant Noomhorm, Chun-Ju Chang, Che-Sheng Wen, Jir-You Wang, Jiun-Liang Chen, Ling-Ming Tseng, Wei-Shone Chen, Jen-Hwey Chiu, Yi-Ming Shyr
ABSTRACT
This study investigated the herb-drug interaction of xanthorrhizol and tamoxifen in human breast cancer cells. Using MCF-7 cell line as an in vitro model, the herb-drug interaction between xanthorrhizol and tamoxifen was measured by MTT assay, luciferase reporter assay, and cell cycle analysis. The effects of xanthorrhizol on growth/autophagy related signaling were determined by immunostaining, western blotting, and real time RT-PCR. Additionally, the in vivo effect of xanthorrhizol and tamoxifen on athymic nude mice implanted with MCF-7 cells was evaluated. When MCF-7 cells were co-treated with tamoxifen and xanthorrhizol, there were no significant changes in terms of cell number, luciferase activity, percentage S-phase cells and LC3-II expression. However, using the MCF-7 implanted nude mice model, it was possible to detect significantly increased tumor volumes, a larger tumor size, and increased protein expression of P38 and P27(Kip1) in the xanthorrhizol + tamoxifen group compared to the tamoxifen-alone group. It can be concluded that while there is no significant herb-drug interaction between xanthorrhizol and tamoxifen in vitro, there is such an interaction in tumor-bearing mice, which provides important information that affects breast cancer treatment translational research.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
SAFC
HEPES
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material