  • Search Results
  • Significant decrease in plasma N-acetyl-seryl-aspartyl-lysyl-proline level in patients with end stage renal disease after kidney transplantation.
Significant decrease in plasma N-acetyl-seryl-aspartyl-lysyl-proline level in patients with end stage renal disease after kidney transplantation.
Biological & pharmaceutical bulletin (2014-06-03)
Yosuke Suzuki, Fumihiko Katagiri, Fuminori Sato, Kanako Fujioka, Yukie Sato, Takashi Fujioka, Yuhki Sato, Hiromitsu Mimata, Hiroki Itoh
ABSTRACT
N-acetyl-seryl-aspartyl-lysyl-proline (AcSDKP) is an endogenous peptide released from its precursor (thymosin-β4) by prolyl oligopeptidase. AcSDKP is a natural inhibitor of pluripotent hematopoietic stem cell proliferation and is normally found in human plasma. AcSDKP has been shown to be a potent angiogenic factor and to suppress renal fibroblast proliferation. Impairment of renal function has been suggested to have a significant impact on plasma AcSDKP level. The aim of this study was to assess whether improvement of renal function after kidney transplantation has an impact on plasma AcSDKP-like immunoreactive substance (IS) level. Fourteen patients with end stage renal disease (ESRD) who were scheduled to undergo the first kidney allograft transplantation were enrolled. Plasma AcSDKP-IS levels were measured before and 3, 7, 10, 14, 21, 30, 60 and 90 d after kidney transplantation. Plasma AcSDKP-IS level decreased significantly from day 3 after kidney transplantation compared to before kidney transplantation. Creatinine clearance increased significantly from day 7 after kidney transplantation. A significant negative correlation was observed between creatinine clearance and plasma AcSDKP-IS level from before transplantation to 90 d after kidney transplantation. Stepwise multiple regression analysis identified creatinine clearance as the only significant independent factor associated with plasma AcSDKP-IS levels. These results suggest that recovery of kidney function after kidney transplantation may lead to a decrease in plasma AcSDKP level in patients with ESRD, and that plasma AcSDKP level may depend largely on renal function.
Sigma-Aldrich
7-Amino-4-methylcoumarin, 99%
Sigma-Aldrich
7-Amino-4-methylcoumarin, Chromophore for substrates
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.