A new ELISA for quantification of brassinosteroids in plants.
Steroids (2014-09-10)
Andrey G Pradko, Raisa P Litvinovskaya, Alina L Sauchuk, Svetlana V Drach, Alexander V Baranovsky, Vladimir N Zhabinskii, Tatyana V Mirantsova, Vladimir A Khripach
ABSTRACT
Starting from (22R,23R)-2α,3α,22,23,26-pentahydroxy-5α-cholestan-6-one 26-hemisuccinate, conjugates of 28-norcastasterone with horse radish peroxidase and bovine serum albumin were prepared. The latter conjugate was injected into rabbits; produced polyclonal antibodies were used to quantitate 6-keto-brassinosteroids. The newly developed analytical system was used in combination with two other immunoenzymatic assays for brassinosteroids to determine individual compounds of this series. In addition, a direct method of brassinosteroid analysis was proposed. It has the advantage of requiring no sample pretreatment steps such as extraction with organic solvents and chromatography.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.7%