  • Search Results
  • Overcoming the susceptibility gap between maternal antibody disappearance and auto-antibody production.
Overcoming the susceptibility gap between maternal antibody disappearance and auto-antibody production.
Vaccine (2014-12-03)
Roni Yosipovich, Elina Aizenshtein, Roy Shadmon, Simcha Krispel, Efrat Shuster, Jacob Pitcovski
ABSTRACT
In the first 10-14 days of a chick's life, protection is conferred by maternal antibodies. Further broiler protection is achieved by active vaccination. However, the high level of maternal antibodies interferes with the induction of an effective immune response by vaccination at a young age. As a result, there is a gap between the reduction in protective maternal antibodies and elevation of self-produced antibodies following active vaccination. The major aim of this study was to test an approach consisting of passive and active vaccination to overcome this gap and to provide continuous resistance to infectious viral diseases during the broiler's growth period. Newcastle disease virus (NDV), which is one of the world's most prevalent infectious diseases of poultry, was tested as a model. Following subcutaneous injection of 18 hemagglutination-inhibiting (HI) units of anti-NDV immunoglobulin Y per 1-day-old chick, protective log2 antibody titers above 4 could be detected to at least 17 days of age. The combination of passive immunization on day 1 of age with attenuated live vaccination on day 10 led to high protective titers throughout the entire growth period, up to 41 days of age. Moreover, the HI titers in the group of birds immunized with the combined vaccination were significantly more homogeneous than those in the group vaccinated only with live virus. Thus, full protection against NDV of all broilers in flock during their entire growth period was achieved by a vaccination regime that combines passive immunization and live vaccination.
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O