  • Search Results
  • In vitro investigation of anticancer, cell-cycle-inhibitory, and apoptosis-inducing effects of diversin, a natural prenylated coumarin, on bladder carcinoma cells.
In vitro investigation of anticancer, cell-cycle-inhibitory, and apoptosis-inducing effects of diversin, a natural prenylated coumarin, on bladder carcinoma cells.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2014-05-31)
Azadeh Haghighitalab, Maryam M Matin, Ahmad Reza Bahrami, Mehrdad Iranshahi, Morvarid Saeinasab, Fereshteh Haghighi
ABSTRACT
Chemotherapy is one of the main strategies for reducing the rate of cancer progression or, in some cases, curing the tumour. Since a great number of chemotherapeutic agents are cytotoxic compounds, i. e. similarly affect normal and neoplastic cells, application of antitumour drugs is preferred in cancer management and therapy. In this study, the cytotoxicity of diversin was evaluated in 5637 cells, a transitional cell carcinoma (TCC) subline (bladder carcinoma), and normal human fibroblast cells using the 3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide (MTT) assay. Chromatin condensation and DNA damage induced by diversin were also determined by means of 4',6-diamidino-2-phenylindole (DAPI) staining and the comet assay, respectively. In addition, the mechanism of action of diversin was studied in more detail by the caspase 3 colourimetric assay and flow cytometry-based cell-cycle analyses (PI staining). Our results revealed that diversin has considerable cytotoxic effects in 5637 cells, but not on HFF3 (human foreskin fibroblast) and HDF1 (human dermal fibroblast) cells. Further studies showed that diversin exerts its cytotoxicity via induction of chromatin condensation, DNA damage, and activation of caspase 3 in 5637 cells. In addition, flow cytometric analyses revealed that 5637 cells are mostly arrested at the G2 phase of the cell cycle in the presence of diversin.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)