  • Search Results
  • Intra-articular delivery of kartogenin-conjugated chitosan nano/microparticles for cartilage regeneration.
Intra-articular delivery of kartogenin-conjugated chitosan nano/microparticles for cartilage regeneration.
Biomaterials (2014-09-23)
Mi Lan Kang, Ji-Yun Ko, Ji Eun Kim, Gun-Il Im
ABSTRACT
We developed an intra-articular (IA) drug delivery system to treat osteoarthritis (OA) that consisted of kartogenin conjugated chitosan (CHI-KGN). Kartogenin, which promotes the selective differentiation of mesenchymal stem cells (MSCs) into chondrocytes, was conjugated with low-molecular-weight chitosan (LMWCS) and medium-molecular-weight chitosan (MMWCS) by covalent coupling of kartogenin to each chitosan using an ethyl(dimethylaminopropyl) carbodiimide (EDC)/N-hydroxysuccinimide (NHS) catalyst. Nanoparticles (NPs, 150 ± 39 nm) or microparticles (MPs, 1.8 ± 0.54 μm) were fabricated from kartogenin conjugated-LMWCS and -MMWCS, respectively, by an ionic gelation using tripolyphosphate (TPP). The in vitro release profiles of kartogenin from the particles showed sustained release for 7 weeks. When the effects of the CHI-KGN NPs or CHI-KGN MPs were evaluated on the in vitro chondrogenic differentiation of human bone marrow MSCs (hBMMSCs), the CHI-KGN NPs and CHI-KGN MPs induced higher expression of chondrogenic markers from cultured hBMMSCs than unconjugated kartogenin. In particular, hBMMSCs treated with CHI-KGN NPs exhibited more distinct chondrogenic properties in the long-term pellet cultures than those treated with CHI-KGN MPs. The in vivo therapeutic effects of CHI-KGN NPs or CHI-KGN MPs were investigated using a surgically-induced OA model in rats. The CHI-KGN MPs showed longer retention time in the knee joint than the CHI-KGN NPs after IA injection in OA rats. The rats treated with CHI-KGN NPs or CHI-KGN MPs by IA injection showed much less degenerative changes than untreated control or rats treated with unconjugated kartogenin. In conclusion, CHI-KGN NPs or CHI-KGN MPs can be useful polymer-drug conjugates as an IA drug delivery system to treat OA.
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
SAFC
L-Proline
Supelco
L-Proline, certified reference material, TraceCERT®
Selenium, pellets, < 5mm, ≥99.999%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Leucine, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.