  • Search Results
  • Technical decision making with higher order structure data: utilization of differential scanning calorimetry to elucidate critical protein structural changes resulting from oxidation.
Technical decision making with higher order structure data: utilization of differential scanning calorimetry to elucidate critical protein structural changes resulting from oxidation.
Journal of pharmaceutical sciences (2015-01-07)
Kelly K Arthur, Nikita Dinh, John P Gabrielson
ABSTRACT
Differential scanning calorimetry (DSC) is a useful tool for monitoring thermal stability of the molecular conformation of proteins. Here, we present an example of the sensitivity of DSC to changes in stability arising from a common chemical degradation pathway, oxidation. This Note is part of a series of industry case studies demonstrating the application of higher order structure data for technical decision making. For this study, six protein products from three structural classes were evaluated at multiple levels of oxidation. For each protein, the melting temperature (Tm ) decreased linearly as a function of oxidation; however, differences in the rate of change in Tm , as well as differences in domain Tm stability were observed across and within structural classes. For one protein, analysis of the impact of oxidation on protein function was also performed. For this protein, DSC was shown to be a leading indicator of decreased antigen binding suggesting a subtle conformation change may be underway that can be detected using DSC prior to any observable impact on product potency. Detectable changes in oxidized methionine by mass spectrometry (MS) occurred at oxidation levels below those with a detectable conformational or functional impact. Therefore, by using MS, DSC, and relative potency methods in concert, the intricate relationship between a primary structural modification, changes in conformational stability, and functional impact can be elucidated.
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Methionine, European Pharmacopoeia (EP) Reference Standard
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis