  • Search Results
  • Sprouting of axonal collaterals after spinal cord injury is prevented by delayed axonal degeneration.
Sprouting of axonal collaterals after spinal cord injury is prevented by delayed axonal degeneration.
Experimental neurology (2014-08-01)
E Collyer, A Catenaccio, D Lemaitre, P Diaz, V Valenzuela, F Bronfman, F A Court
ABSTRACT
After an incomplete spinal cord injury (SCI), partial recovery of locomotion is accomplished with time. Previous studies have established a functional link between extension of axon collaterals from spared spinal tracts and locomotor recovery after SCI, but the tissular signals triggering collateral sprouting have not been identified. Here, we investigated whether axonal degeneration after SCI contributes to the sprouting of collaterals from axons spared after injury. To this end, we evaluated collateral sprouting from BDA-labeled uninjured corticospinal axons after spinal cord hemisection (SCI(H)) in wild type (WT) mouse and Wld(S) mouse strains, which shows a significant delay in Wallerian degeneration after injury. After SCI(H), spared fibers of WT mice extend collateral sprouts to both intact and denervated sides of the spinal cord distant from the injury site. On the contrary, in the Wld(S) mice collateral sprouting from spared fibers was greatly reduced after SCI(H). Consistent with a role for collateral sprouting in functional recovery after SCI, locomotor recovery after SCI(H) was impaired in Wld(S) mice compared to WT animals. In conclusion, our results identify axonal degeneration as one of the triggers for collateral sprouting from the contralesional uninjured fibers after an SCI(H). These results open the path for identifying molecular signals associated with tissular changes after SCI that promotes collateral sprouting and functional recovery.
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Supelco
(±)-Propylene oxide, analytical standard
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, affinity isolated antibody