  • Search Results
  • Nuclear erythroid 2 p45-related factor-2 Nrf2 ameliorates cigarette smoking-induced mucus overproduction in airway epithelium and mouse lungs.
Nuclear erythroid 2 p45-related factor-2 Nrf2 ameliorates cigarette smoking-induced mucus overproduction in airway epithelium and mouse lungs.
Microbes and infection (2014-09-23)
Ying-Hua Ying, Xiao-Ping Lin, Hong-bin Zhou, Yin-fang Wu, Fu-gui Yan, Wen Hua, Li-Xia Xia, Zhang-wei Qiu, Zhi-Hua Chen, Wen Li, Hua-Hao Shen
ABSTRACT
Nuclear erythroid 2 p45-related factor-2 (Nrf2) is known to play important roles in airway disorders, whereas little has been investigated about its direct role in airway mucus hypersecretion. The aim of this study is to determine whether this factor could protect pulmonary epithelium and mouse airway from cigarette-induced mucus overproduction. Using genetic approaches, the role of Nrf2 on cigarette smoking extracts (CSE) induced MUC5AC expression was investigated in lung A549 cells. Nrf2 deficiency mice were smoked for various periods, and the airway inflammation and mucus production was characterized. Acute smoking exposure induced expression of MUC5AC and Nrf2 in both A549 cells and mouse lungs. Genetic ablation of Nrf2 augmented, whereas overexpression of this molecule ameliorated CSE-induced expression of MUC5AC. Nrf2 knockout mice, after exposure to cigarette smoking, displayed enhanced airway inflammation and mucus production. Nrf2 negatively regulated smoking-induced mucus production in vitro and in vivo, suggesting therapeutic potentials of this factor in airway diseases with hypersecreted mucus.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material