  • Search Results
  • Interference of Ca²⁺ with the proliferation of SCCOHT-1 and ovarian adenocarcinoma cells.
Interference of Ca²⁺ with the proliferation of SCCOHT-1 and ovarian adenocarcinoma cells.
International journal of oncology (2014-06-28)
Anna Otte, Finn Rauprich, Juliane von der Ohe, Peter Hillemanns, Ralf Hass
ABSTRACT
A recently established cellular model for the rare small cell carcinoma of the ovary hypercalcemic type (SCCOHT-1) was characterized in comparison to ovarian adenocarcinoma cells (NIH:OVCAR-3 and SK-OV-3). The different cancer populations exhibited a common sensitivity in acidic pH milieu and a continuous proliferation in alkaline medium of pH 8.0-9.0. In the presence of elevated Ca2+ concentrations, the ovarian cancer cells demonstrated a progressively reduced proliferation within 72 h in contrast to other tumor types such as breast cancer cells. This significant growth inhibition was calcium-specific since the proliferation was unaffected after culture of the ovarian cancer cells in the presence of similar concentrations of other cations. The Ca2+ effects on the ovarian cancer cells were associated with marked differences in the activation of intracellular signaling pathways including enhanced phosphorylation of the p42/44 MAP kinase (Thr202/Tyr204). Further analysis of the signaling pathway revealed a significantly enhanced Ca2+-dependent and p42/44 MAP kinase activation-mediated prostaglandin E2 (PGE2) production in SK-OV-3 and SCCOHT-1 and to a lesser extent in NIH:OVCAR-3 cells. Vice versa, exogenous PGE2 did not affect the proliferative capacity of the ovarian cancer cells and inhibition of the Ca2+-mediated MAP kinase activation did not abolish the Ca2+-mediated cytotoxicity. Collectively, these data suggest that multiple pathways are activated by exogenous Ca2+ in the different ovarian cancer cells, including a specific MAP kinase signaling cascade with subsequent PGE2 production and a parallel pathway for the induction of cell death.
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
PD 98,059, solid