  • Search Results
  • In vitro and in vivo characterisation of Lu AF64280, a novel, brain penetrant phosphodiesterase (PDE) 2A inhibitor: potential relevance to cognitive deficits in schizophrenia.
In vitro and in vivo characterisation of Lu AF64280, a novel, brain penetrant phosphodiesterase (PDE) 2A inhibitor: potential relevance to cognitive deficits in schizophrenia.
Psychopharmacology (2014-03-01)
John P Redrobe, Morten Jørgensen, Claus T Christoffersen, Liliana P Montezinho, Jesper F Bastlund, Martin Carnerup, Christoffer Bundgaard, Linda Lerdrup, Niels Plath
ABSTRACT
Here, we present the pharmacological characterisation of Lu AF64280, a novel, selective, brain penetrant phosphodiesterase (PDE) 2A inhibitor, in in vitro/in vivo assays indicative of PDE2A inhibition, and in vivo models/assays relevant to cognitive processing or antipsychotic-like activity. The in vitro selectivity of Lu AF64280 was determined against a panel of PDE enzymes and 3',5'-cyclic guanosine monophosphate (cGMP) levels in the hippocampus were determined using in vivo microdialysis. Lu AF64280 potently inhibited hPDE2A (Ki = 20 nM), 50-fold above moderate inhibition of both hPDE9A (Ki = 1,000 nM) and hPDE10A (Ki = 1,800 nM), and displayed a >250-fold selectivity over all other full-length human recombinant PDE family members (Ki above 5,000 nM). Lu AF64280 (20 mg/kg) significantly increased cGMP levels in the hippocampus (p < 0.01 versus vehicle-treated mice), attenuated sub-chronic phencyclidine-induced deficits in novel object exploration in rats (10 mg/kg, p < 0.001 versus vehicle-treated), blocked early postnatal phencyclidine-induced deficits in the intradimensional/extradimensional shift task in rats (1 and 10 mg/kg, p < 0.001 versus vehicle-treated) and attenuated spontaneous P20-N40 auditory gating deficits in DBA/2 mice (20 mg/kg, p < 0.05 versus vehicle-treated). In contrast, Lu AF64280 failed to attenuate phencyclidine-induced hyperactivity in mice, and was devoid of antipsychotic-like activity in the conditioned avoidance response paradigm in rats, at any dose tested. Lu AF64280 represents a novel tool compound for selective PDE2A inhibition that substantiates a critical role of this enzyme in cognitive processes under normal and pathological conditions.
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate sodium salt, ≥99% (HPLC), powder
Sevoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
USP
Haloperidol, United States Pharmacopeia (USP) Reference Standard