  • Search Results
  • Dendritic cells transfected with hepatocellular carcinoma (HCC) total RNA induce specific immune responses against HCC in vitro and in vivo.
Dendritic cells transfected with hepatocellular carcinoma (HCC) total RNA induce specific immune responses against HCC in vitro and in vivo.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico (2013-12-18)
B H Xie, J Y Yang, H P Li, B Zhang, W Chen, B Zhou, B G Peng, L J Liang, Q He
ABSTRACT
Immunotherapy is an effective method for preventing metastasis and recurrence of carcinoma. Hepatocellular carcinoma (HCC) is a common malignancy with a high rate of recurrence, and has not successfully been introduced to immunotherapy. Peripheral blood mononuclear cells were isolated from whole blood of HCC patients and stimulated to transform into dendritic cells (DCs). These DCs were then transfected with RNA extracted from HepG-2 hepatoma cells to induce expression of specific antigens. The transfected DCs stimulated T lymphocytes to produce cytotoxic T lymphocytes, which specifically attacked HepG-2 cells. Injection of T lymphocytes from HCC patients and transfected DCs into severe combined immunodeficiency mice limited the growth of HepG-2 tumors. A specific immune response against hepatoma can be generated in vivo by administering DCs transfected with RNA from a specific tumor. This method may have therapeutic application in humans to reduce recurrence of HCC.
Sigma-Aldrich
IL-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Macrophage Colony-Stimulating Factor human, M-CSF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
IL-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.