  • Search Results
  • Enhanced spectrophotometric determination of Losartan potassium based on its physicochemical interaction with cationic surfactant.
Enhanced spectrophotometric determination of Losartan potassium based on its physicochemical interaction with cationic surfactant.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2014-10-16)
Laila Abdel-Fattah, Lobna Abdel-Aziz, Mariam Gaied
ABSTRACT
In this study, a simple and sensitive spectrophotometric method was developed for determination of Losartan potassium (LST K), an angiotensin-II receptor (type AT1) antagonist, in presence of cationic surfactant cetyltrimethylammonium bromide (CTAB). The physicochemical interaction of LST K with CTAB was investigated. The effect of cationic micelles on the spectroscopic and acid-base properties of LST K was studied at pH 7.4. The binding constant (Kb) and the partition coefficient (Kx) of LST K-CTAB were 1.62×10(5) M(-1) and 1.38×10(5); respectively. The binding of LST K to CTAB micelles implied a shift in drug acidity constant (ΔpKa=0.422). The developed method is linear over the range 0.5-28 μg mL(-1). The accuracy was evaluated and was found to be 99.79±0.509% and the relative standard deviation for intraday and interday precision was 0.821 and 0.963; respectively. The method was successfully applied to determine LST K in pharmaceutical formulations.
Product Description
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Supelco
Citric acid, analytical standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%