  • Search Results
  • Hyperglycemic arterial disturbed flow niche as an in vitro model of atherosclerosis.
Hyperglycemic arterial disturbed flow niche as an in vitro model of atherosclerosis.
Analytical chemistry (2014-10-04)
Phani K Patibandla, Aaron J Rogers, Guruprasad A Giridharan, Manuel A Pallero, Joanne E Murphy-Ullrich, Palaniappan Sethu
ABSTRACT
Type 2 diabetes significantly elevates the risk of cardiovascular disease. This can be largely attributed to the adverse effects of hyperglycemic conditions on normal endothelial cell (EC) function. ECs in both large and small vessels are influenced by hyperglycemic conditions, which increase susceptibility to EC dysfunction and atherosclerotic lesion formation. Fluid shear stress and flow patterns play an essential role in atherogenesis: lesions form only at locations where fluid flow behavior can be classified as "disturbed flow" (i.e., low shear stress recirculation and/or retrograde flow). Since regions of disturbed flow are the focal points of atherosclerotic cardiovascular disease, we hypothesized that the combinatorial effects of high glucose and disturbed flow conditions elicit significantly different responses from ECs than high glucose alone. To validate our hypothesis, we used our endothelial cell culture model (ECCM) to establish vascular niches associated with "normal" and "disturbed" flow conditions typically seen in vivo along with physiological pressure and stretch. We subjected human aortic endothelial cells (HAECs) to hyperglycemic conditions under both "normal" and "disturbed" flow. Our results confirm significant and quantifiable differences in phenotypic and functional markers between cells cultured under conditions of "normal" and "disturbed flow" under hyperglycemic conditions suggesting that elevated glucose in conjunction with "disturbed" flow conditions results in significantly higher level of EC dysfunction. The ECCM can therefore be used as a physiologically relevant model to study early stage hyperglycemia induced atherosclerosis for basic research, drug discovery, and screening and toxicity studies.
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-eNOS (pSer1176) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-phospho-eNOS (pThr494) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-eNOS (pSer615) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-NOS3 (pSer1177) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6D6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2H1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6G6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Rac antibody produced in mouse, clone 23A8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Rabbit anti-AKT1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.