  • Search Results
  • The influence of intestinal lymphatic transport on the systemic exposure and brain deposition of a novel highly lipophilic compound with structural similarity to cholesterol.
The influence of intestinal lymphatic transport on the systemic exposure and brain deposition of a novel highly lipophilic compound with structural similarity to cholesterol.
The Journal of pharmacy and pharmacology (2014-05-14)
Suzanne M Caliph, Fried W Faassen, Christopher J H Porter
ABSTRACT
To assess the role of intestinal lymphatic transport in the oral bioavailability and brain deposition of a highly lipophilic, centrally acting drug candidate (Org 49209) in comparison to cholesterol, a close structural analogue. The intestinal lymphatic transport of Org 49209 and cholesterol was assessed in lymph-cannulated anaesthetised rats and total bioavailability evaluated in non-lymph-cannulated animals. Parallel groups were employed to examine the brain deposition of Org 49209 after intraduodenal and intraperitoneal administrations. The contribution of intestinal lymphatic transport to total bioavailability was similar for Org 49209 and cholesterol (approximately 40% of the absorbed dose). However, the oral bioavailability of Org 49209 was significantly (fourfold) lower than cholesterol. Brain deposition of Org 49209 was similar after intraduodenal and intraperitoneal administration. Systemic exposure, however, was higher after intraduodenal administration and brain-to-plasma ratios were therefore reduced. The oral bioavailability of Org 49209 was significantly lower than that of its structural analogue cholesterol; however, intestinal lymphatic transport played a similar role in oral bioavailability for both compounds. Brain to plasma ratios were lower after intraduodenal versus intraperitoneal administration, suggesting that drug association with intestinal lymph lipoproteins may limit central nervous system access for highly lipophilic drugs.
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, natural, FCC
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, Selectophore, ≥99.0%
Oleic acid, European Pharmacopoeia (EP) Reference Standard