  • Search Results
  • Compared to controls, patients with ruptured aneurysm and surgical intervention show increase in symptoms of depression and lower cognitive performance, but their objective sleep is not affected.
Compared to controls, patients with ruptured aneurysm and surgical intervention show increase in symptoms of depression and lower cognitive performance, but their objective sleep is not affected.
The world journal of biological psychiatry : the official journal of the World Federation of Societies of Biological Psychiatry (2014-02-26)
Serge Brand, Stefan Zimmerer, Nadeem Kalak, Sandra Von Planta, Katja Schwenzer-Zimmerer, Andreas Albert Müller, Hans-Florian Zeilhofer, Edith Holsboer-Trachsler
ABSTRACT
Patients with aneurysmal subarachnoid haemorrhage (aSAH) have impaired sleep and cognitive performance together with more difficulties in social and everyday life. Hypocortisolism has also been reported. However, a study assessing all dimensions between aSAH severity, objective and subjective sleep, cortisol secretion, cognitive performance and social and everyday life has not so far been performed. The aim of the present study was therefore two-fold: (1) to assess, in a sample of patients with aSAH, objective and subjective sleep, cognitive functioning, social skills and cortisol secretion concurrently, and (2) to compare patients on these variables with a control group. Twenty-one patients (17 females; mean age: 58.80 years) with ruptured aneurysm and surgical intervention and 21 (14 females; mean age: 58.90 years) age- and gender-matched controls took part in the study. Assessments covered objective sleep-EGG recordings, subjective sleep, salivary cortisol analysis, and psychological functioning including memory performance, mood, and emotion recognition. Compared to healthy controls, patients had lower scores for verbal memory performance and emotion recognition; they also reported more marked depressive symptoms and complained of poor sleep. However, no differences were found for objective sleep or cortisol secretion. Subjective and objective sleep, cortisol secretion and psychological functioning were unrelated. Findings indicate that patients with aSAH face psychological rather than physiological issues.
Product Description
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard