Sigma-Aldrich
Anti-NFKB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-JNK1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-NF-κB p105 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-NFKB1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2H1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6G6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-MAPK8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Mnk1 (pThr385) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-NFKB1 (Ab-927) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-NFKB1 (Ab-907) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-NFKB1 (Ab-932) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pThr450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-RELA (pSer529) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MAPK3 (Ab-204) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MAPK3 (Ab-202) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Monoclonal Anti-NF-κB antibody produced in mouse, clone NF-12, ascites fluid
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-MAPK3 antibody produced in mouse, clone 3C9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-NFKB1 antibody produced in mouse, clone 2E6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution