  • Search Results
  • Radiation induces phosphorylation of STAT3 in a dose- and time-dependent manner.
Radiation induces phosphorylation of STAT3 in a dose- and time-dependent manner.
Asian Pacific journal of cancer prevention : APJCP (2014-08-16)
Ling Gao, Feng-Sheng Li, Xiao-Hua Chen, Qiao-Wei Liu, Jiang-Bin Feng, Qing-Jie Liu, Xu Su
ABSTRACT
We have reported the radiation could activate STAT3, which subsequently promotes the invasion of A549 cells. We here explored the dose- and time-response of STAT3 to radiation and the effect of radiation on upstream signaling molecules. A549 cells were irradiated with different doses of γ-rays. The expression of and nucleus translocation of p-STAT3 in A549 cells were detected by immunoblotting and immunofluorescence, respectively. The level of phosphorylated EGFR was also assessed by immunoblotting, and IL-6 expression was detected by real time PCR and ELISA. Radiation promoted the phosphorylation of STAT3 at Y705 in a dose- and time-dependent manner and nuclear translocation. The level of phosphorylated EGFR in A549 cells increased after radiation. In additional, the mRNA and protein levels of IL-6 in A549 cells were also up regulated by radiation. STAT3 is activated by radiation in a dose-and time-dependent manner, probably due to radiation-induced activation of EGFR or secretion of IL-6 in A549 cells.
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants