  • Search Results
  • The positive is inside the negative: HER2-negative tumors can express the HER2 intracellular domain and present a HER2-positive phenotype.
The positive is inside the negative: HER2-negative tumors can express the HER2 intracellular domain and present a HER2-positive phenotype.
Cancer letters (2014-12-02)
Carolina Panis, Luciana Pizzatti, Stephany Corrêa, Renata Binato, Gabriela Ferreira Lemos, Ana Cristina da Silva do Amaral Herrera, Teresa Fernandes Seixas, Rubens Cecchini, Eliana Abdelhay
ABSTRACT
Overexpression of human epithelial growth factor receptor 2 (HER2) is a poor prognostic factor in breast cancer. HER2 is a transmembrane receptor comprising an extracellular domain (ECD), a single transmembrane domain, and an intracellular domain (ICD) with tyrosine-kinase activity. Receptor dimerization triggers pivotal effector pathways in cancer, such as phosphatidylinositol 3-kinase (PI3K) signaling. Currently, screening of HER2 in breast tumors for prognostic and therapeutic purposes involves immunohistochemical (IHC) phenotyping for the ECD, in which tumors with IHC scores below 2+ are reported as HER2-negative. We used a label-free liquid chromatography-mass spectrometry (LC-MS) proteomic approach to compare plasma samples from patients with HER2-positive breast tumors and patients with HER2-negative tumors. Patients with HER2-negative tumors expressed higher circulating levels of calpain-10 than patients with HER2-positive tumors. Calpains cleave HER2, releasing its ECD and transforming phenotypically positive tumors into phenotypically negative tumors. Therefore, we investigated the expression of the ICD in HER2-negative samples that overexpressed calpain-10. We found that 16% of HER2-negative tumors were positive for HER2-ICD, which was associated with circulating HER2-ECD. HER2 gene amplification was also observed in some HER2-negative tumors. Positive staining for the PI3K pathway was observed in the HER2-negative, ICD-positive tumors, similar to the HER2-positive cohort. Microarray analysis revealed that HER2-negative, ICD-positive samples clustered between HER2-positive tumors and triple-negative tumors. Survival analysis revealed that outcome in women with HER2-negative, ICD-positive tumors was better than in women bearing HER2-negative, ICD-negative (triple negative) tumors but was quite similar to HER2-positive tumors and worse than women with luminal A tumors. Moreover, in vitro analyses revealed that MDA-MB 231, a triple negative cell line, possesses calpain-10 and HER2-p-ICD up-regulation and blockage of calpain-10 activity promoted an increase in HER2-p-ICD and p-AKT levels, suggesting an increase in these pathways signaling. These data indicate that HER2-negative tumors with HER2-ICD positivity exhibit clinical behavior closer to that of HER2-positive tumors. This indicates a need for HER2-ICD screening when determining the molecular profile of breast tumors. These findings further indicate that lapatinib should be investigated as a target therapy for HER2-ICD-positive breast tumors.
Product Description
Supelco
Hematoxylin
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Supelco
Progesterone, VETRANAL®, analytical standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard