  • Search Results
  • Increased CD4+CD25+ regulatory T cells correlate with poor short-term outcomes in hepatitis B virus-related acute-on-chronic liver failure patients.
Increased CD4+CD25+ regulatory T cells correlate with poor short-term outcomes in hepatitis B virus-related acute-on-chronic liver failure patients.
Journal of microbiology, immunology, and infection = Wei mian yu gan ran za zhi (2014-01-07)
Chuan Shen, Wen-Zhao Yan, Cai-Yan Zhao, Hong-Hao Che, Xiao-Yu Liu, Zhen-Zhong Liu, Ya-Dong Wang, Wei Wang, Meng Li, Jian Gao
ABSTRACT
The roles of CD4(+)CD25(+) regulatory T cells (Treg) in chronicity of hepatitis B virus (HBV) infection have been confirmed. We aimed to explore alteration of Treg in patients with HBV-related acute-on-chronic liver failure (ACLF). Thirty-two HBV-related ACLF patients, 44 chronic hepatitis B patients, and 41 healthy controls were recruited. We detected frequencies of peripheral Treg and intrahepatic forkhead winged helix transcription factor (Foxp3)(+) cells. Inhibitory activity of Treg was assessed by functional suppression assays. Serum interferon-γ and interleukin-10 were also determined. Peripheral Treg and intrahepatic Foxp3(+) cells were more markedly increased in ACLF than chronic hepatitis B and controls (all p < 0.001), and the Foxp3(+) cells located predominantly in the portal areas. The Treg frequency was positively correlated with HBV DNA load, international normalized ratio, model of end stage liver disease score, and serum interleukin-10 level in ACLF patients. Functional assays in vitro demonstrated that ACLF patients exhibited higher suppressive effects of Treg on proliferations of autologous CD4(+)CD25(-) T cells than controls. On logistic regression, prolonged international normalized ratio and higher peripheral Treg frequency predicted 30-day survival of ACLF. The patients with HBV-related ACLF exhibit increased amounts of Treg, of which redistribution from periphery to liver seems to modulate liver inflammation. Higher Treg amounts are associated with more severe liver disease in ACLF, and its level in combination with international normalized ratio may assist prediction of short-term outcomes of HBV-related ACLF.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer