  • Search Results
  • Cardioprotective and cardiotoxic effects of quercetin and two of its in vivo metabolites on differentiated h9c2 cardiomyocytes.
Cardioprotective and cardiotoxic effects of quercetin and two of its in vivo metabolites on differentiated h9c2 cardiomyocytes.
Basic & clinical pharmacology & toxicology (2014-09-10)
James Daubney, Philip L Bonner, Alan J Hargreaves, John M Dickenson
ABSTRACT
Whilst mitotic rat embryonic cardiomyoblast-derived H9c2 cells have been widely used as a model system to study the protective mechanisms associated with flavonoids, they are not fully differentiated cardiac cells. Hence, the aim of this study was to investigate the cardioprotective and cardiotoxic actions of quercetin and two of its major in vivo metabolites, quercetin 3-glucuronide and 3'-O-methyl quercetin, using differentiated H9c2 cells. The differentiated cardiomyocyte-like phenotype was confirmed by monitoring expression of cardiac troponin 1 after 7 days of culture in reduced serum medium containing 10 nM all-trans retinoic acid. Quercetin-induced cardiotoxicity was assessed by monitoring MTT reduction, lactate dehydrogenase (LDH) release, caspase 3 activity and reactive oxygen species production after prolonged flavonoid exposure (72 hr). Cardiotoxicity was observed with quercetin and 3'-O-methyl quercetin, but not quercetin 3-glucuronide. Cardioprotection was assessed by pre-treating differentiated H9c2 cells with quercetin or its metabolites for 24 hr prior to 2-hr exposure to 600 μM H2 O2, after which oxidative stress-induced cell damage was assessed by measuring MTT reduction and LDH release. Cardioprotection was observed with quercetin and 3'-O-methyl quercetin, but not with quercetin 3-glucuronide. Quercetin attenuated H2 O2 -induced activation of ERK1/2, PKB, p38 MAPK and JNK, but inhibitors of these kinases did not modulate quercetin-induced protection or H2 O2 -induced cell death. In summary, quercetin triggers cardioprotection against oxidative stress-induced cell death and cardiotoxicity after prolonged exposure. Further studies are required to investigate the complex interplay between the numerous signalling pathways that are modulated by quercetin and which may contribute to the cardioprotective and cardiotoxic effects of this important flavonoid.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%