  • Search Results
  • Specific regions display altered grey matter volume in μ-opioid receptor knockout mice: MRI voxel-based morphometry.
Specific regions display altered grey matter volume in μ-opioid receptor knockout mice: MRI voxel-based morphometry.
British journal of pharmacology (2014-06-11)
Kazumasu Sasaki, Akira Sumiyoshi, Hiroi Nonaka, Yoshiyuki Kasahara, Kazutaka Ikeda, F Scott Hall, George R Uhl, Masahiko Watanabe, Ryuta Kawashima, Ichiro Sora
ABSTRACT
μ Opioid receptor knockout (MOP-KO) mice display several behavioural differences from wild-type (WT) littermates including differential responses to nociceptive stimuli. Brain structural changes have been tied to behavioural alterations noted in transgenic mice with targeting of different genes. Hence, we assess the brain structure of MOP-KO mice. Magnetic resonance imaging (MRI) voxel-based morphometry (VBM) and histological methods were used to identify structural differences between extensively backcrossed MOP-KO mice and WT mice. MOP-KO mice displayed robust increases in regional grey matter volume in olfactory bulb, several hypothalamic nuclei, periaqueductal grey (PAG) and several cerebellar areas, most confirmed by VBM analysis. The largest increases in grey matter volume were detected in the glomerular layer of the olfactory bulb, arcuate nucleus of hypothalamus, ventrolateral PAG (VLPAG) and cerebellar regions including paramedian and cerebellar lobules. Histological analyses confirm several of these results, with increased VLPAG cell numbers and increased thickness of the olfactory bulb granule cell layer and cerebellar molecular and granular cell layers. MOP deletion causes previously undescribed structural changes in specific brain regions, but not in all regions with high MOP receptor densities (e.g. thalamus, nucleus accumbens) or that exhibit adult neurogenesis (e.g. hippocampus). Volume differences in hypothalamus and PAG may reflect behavioural changes including hyperalgesia. Although the precise relationship between volume change and MOP receptor deletion was not determined from this study alone, these findings suggest that levels of MOP receptor expression may influence a broader range of neural structure and function in humans than previously supposed. This article is part of a themed section on Opioids: New Pathways to Functional Selectivity. To view the other articles in this section visit http://dx.doi.org/10.1111/bph.2015.172.issue-2.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Lithium carbonate, ACS reagent, ≥99.0%
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Lithium carbonate, 99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium carbonate, puriss. p.a., ACS reagent, reagent (for microscopy), ≥99.0% (T)
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Lithium carbonate, 99.997% trace metals basis
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid