  • Search Results
  • Cross-species antibody microarray interrogation identifies a 3-protein panel of plasma biomarkers for early diagnosis of pancreas cancer.
Cross-species antibody microarray interrogation identifies a 3-protein panel of plasma biomarkers for early diagnosis of pancreas cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-01-16)
Justin E Mirus, Yuzheng Zhang, Christopher I Li, Anna E Lokshin, Ross L Prentice, Sunil R Hingorani, Paul D Lampe
ABSTRACT
Pancreatic ductal adenocarcinoma (PDA) is the fourth leading cause of cancer-related death in the United States, and its incidence is on the rise. Advanced disease is nearly uniformly lethal, emphasizing the need to identify PDA at its earliest stages. To discover early biomarkers of PDA, we evaluated the circulating proteome in murine preinvasive and invasive plasma samples and human prediagnostic and diagnostic samples. Using a customized antibody microarray platform containing >4,000 features, we interrogated plasma samples spanning preinvasive and invasive disease from a highly faithful mouse model of PDA. In parallel, we mined prediagnostic plasma from women in the Women's Health Initiative (WHI) who would later succumb to PDA together with matched, cancer-free control samples. Samples collected after an establishing diagnosis of PDA were also interrogated to further validate markers. We identified ERBB2 and TNC in our cross-species analyses, and multiple antibodies identified ESR1 in prediagnostic plasma from people that succumb to PDA. This 3-marker panel had an AUC of 0.86 (95% confidence interval [CI], 0.76-0.96) for the diagnostic cohort that increased to 0.97 (95% CI, 0.92-1.0) with CA19-9 included. The 3-marker panel also had an AUC of 0.68 (95% CI, 0.58-0.77) for the prediagnostic cohort. We identified potential disease detection markers in plasma up to 4 years before death from PDA with superior performance to CA19-9. These markers might be especially useful in high-risk cohorts to diagnose early, resectable disease, particularly in patients that do not produce CA19-9.
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O