  • Search Results
  • A mutated anti-CA19-9 scFv-Fc for positron emission tomography of human pancreatic cancer xenografts.
A mutated anti-CA19-9 scFv-Fc for positron emission tomography of human pancreatic cancer xenografts.
Molecular imaging and biology (2014-04-03)
Matthew M Rochefort, Mark D Girgis, Scott M Knowles, Jacob S Ankeny, Felix Salazar, Anna M Wu, James S Tomlinson
ABSTRACT
Intact antibodies have a long serum persistence resulting in high background signal that inhibits their direct translation as imaging agents. Engineering of antibody fragments through the introduction of mutations in the fragment crystallizable (Fc) region can dramatically reduce serum persistence. We sought to develop a Fc-mutated, anti-CA19-9 antibody fragment (anti-CA 19-9 scFv-Fc H310A) to provide micro-positron emission tomography (microPET) imaging of pancreatic cancer xenografts. The anti-CA19-9 scFv-Fc H310A was successfully expressed and purified. Biochemical characterization included size exclusion chromatography, sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE), Western blot, and flow cytometry. The antibody fragment was labeled with iodine-124 ((124)I) and injected into mice containing human pancreatic cancer xenografts. MicroPET/CT images were then obtained. Blood, organ, and tumor radioactivity was measured and expressed as the percent of injected dose per gram of tissue (%ID/g). Biochemical characterization was consistent with the creation of a 105 kD dimer containing a human Fc region. Flow cytometry demonstrated antigen-specific binding, and cell-based ELISA further established a dissociation constant (K D) of 10.7 nM. (124)I-labeled scFv-Fc H310A localized to the antigen-positive tumor xenografts as detected by microPET. Objective confirmation of targeting was demonstrated by higher %ID/g in the antigen-positive tumor compared to the blood, antigen-negative tumor, and liver. We successfully engineered and produced an anti-CA19-9 scFv-Fc H310A antibody fragment that retains similar affinity when compared to the parental intact murine antibody. Additionally, our engineered and mutated fragment exhibited antigen-specific microPET imaging of both subcutaneous and orthotopic pancreatic cancer xenografts at early time points secondary to decreased serum half-life.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules