  • Search Results
  • Autophagic cell death of human hepatoma G2 cells mediated by procyanidins from Castanea mollissima Bl. Shell-induced reactive oxygen species generation.
Autophagic cell death of human hepatoma G2 cells mediated by procyanidins from Castanea mollissima Bl. Shell-induced reactive oxygen species generation.
Chemico-biological interactions (2014-10-08)
Yuqing Duan, Jiajia Ke, Haihui Zhang, Yuanqing He, Guibo Sun, Xiaobo Sun
ABSTRACT
The autophagy of human hepatoma G2 (HepG2) cells induced by procyanidins from chestnut (Castanea mollissima Bl.) shell (CSPCs) was investigated, and the inherent relationship between autophagic levels and reactive oxygen species (ROS) generation was studied. The results showed that CSPCs induced HepG2 cell death in a time- and concentration-dependent manner, increased the accumulation of autophagolysosomes and microtubule-associated proteins light chain 3-II (LC3-II, a marker of autophagy). However, these phenomena were not observed in the group pretreated with the autophagy inhibitor 3-MA, suggesting that CSPCs induced HepG2 cell autophagy. Furthermore, we found that CSPCs triggered ROS generation in cells, while the levels of ROS decreased in the N-acetylcysteine (Nac) co-treatment, revealing that CSPCs-mediated autophagy was partly blocked by Nac. In addition, treatment with CSPCs decreased the mitochondrial membrane potential of HepG2 cells. These results suggested CSPCs could trigger autophagy via ROS generation, which may be associated with the mitochondria-dependent signaling way.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2′,7′-Dichlorofluorescein diacetate, BioReagent, suitable for fluorescence, ≥95% (HPLC)
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
JC-1, solid
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard