  • Search Results
  • Synergistic antitumor activity of withaferin A combined with oxaliplatin triggers reactive oxygen species-mediated inactivation of the PI3K/AKT pathway in human pancreatic cancer cells.
Synergistic antitumor activity of withaferin A combined with oxaliplatin triggers reactive oxygen species-mediated inactivation of the PI3K/AKT pathway in human pancreatic cancer cells.
Cancer letters (2014-12-03)
Xu Li, Feng Zhu, Jianxin Jiang, Chengyi Sun, Xin Wang, Ming Shen, Rui Tian, Chengjian Shi, Meng Xu, Feng Peng, Xingjun Guo, Min Wang, Renyi Qin
ABSTRACT
Application of oxaliplatin for the treatment of pancreatic cancer (PC) is restricted owing to its toxic side effects and drug resistance. We investigated how withaferin A (WA), a bioactive component isolated from the medicinal plant Withania somnifera, acts synergistically with oxaliplatin on human PC in vitro and in vivo. We found that WA enhanced oxaliplatin-induced growth suppression and apoptosis in PC cells dramatically through a mechanism involving mitochondrial dysfunction and inactivation of the PI3K/AKT pathway. Combination treatment resulted in significant accumulation of intracellular reactive oxygen species (ROS). Pretreatment of cells with the ROS scavenger N-acetylcysteine completely blocked the apoptosis induced by combination treatment, and recovered expression of AKT inactivation, which revealed the important role of ROS in apoptosis and AKT regulation. In vivo, combination therapy showed the strongest anti-tumor effects compared with single agents, without obvious additional toxicity. These results support the notion that combination treatment with oxaliplatin and WA could facilitate development of an effective strategy for PC treatment.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Cyclosporin A, 98.5-101.5% (on dried basis)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)