  • Search Results
  • Disrupted MEK/ERK signaling in the medial orbital cortex and dorsal endopiriform nuclei of the prefrontal cortex in a chronic restraint stress mouse model of depression.
Disrupted MEK/ERK signaling in the medial orbital cortex and dorsal endopiriform nuclei of the prefrontal cortex in a chronic restraint stress mouse model of depression.
Neuroscience letters (2014-08-15)
Yea-Hyun Leem, Sang-Sun Yoon, Yu-Han Kim, Sangmee Ahn Jo
ABSTRACT
Depression is one of the most prevalent mental illnesses, and causes a constant feeling of sadness and lose of interest, which often leads to suicide. Evidence suggests that depression is associated with aberrant MEK/ERK signaling. However, studies on MEK/ERK signaling in depression have only been done in a few brain regions, such as the hippocampus and mesolimbic reward pathways. Recent studies also implicate the involvement of the prefrontal cortex in depression. Thus, we examined the changes in MEK/ERK signaling in subregions of the prefrontal cortex of C57BL/6 mice by immunohistochemistry using phospho-MEK1/2 (Ser 217/221) and ERK1/2 (Thr202/Tyr204) antibodies. Mice were subjected to 21 consecutive days of restraint stress (for 2h daily), and depression-like behavior was evaluated using a sociability test and tail suspension test. The antidepressant, imipramine (20mg/kg) was injected intraperitoneally 30min before restraint stress exposure. Chronic/repeated restraint stress produced depressive-like behavior, such as increased social avoidance in the social interaction test, and enhanced immobility time in the tail suspension test. This depressive behavior was ameliorated by imipramine. The behavioral changes well corresponded to a decrease in MEK/ERK immunoreactivity in the medial orbital (MO) cortex and dorsal endopiriform nuclei (DEn), which was averted by imipramine, but not in cingulate, prelimbic, infralimbic, and motor cortex. These results suggest that MEK/ERK signaling is disrupted in the DEn and MO subregions of the prefrontal cortex in the depressive phenotype, and that blocking a decrease in activated MEK/ERK is inherent to the antidepressant imipramine response.
Sigma-Aldrich
Anti-phospho-MAP2K1 (pThr386) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MAP2K1 (pSer221) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MAP2K1 (pThr291) antibody produced in rabbit, affinity isolated antibody