  • Search Results
  • Hypometabolism and hypothermia in the rat model of endotoxic shock: independence of circulatory hypoxia.
Hypometabolism and hypothermia in the rat model of endotoxic shock: independence of circulatory hypoxia.
The Journal of physiology (2014-06-22)
Joshua J Corrigan, Monique T Fonseca, Elizabeth A Flatow, Kevin Lewis, Alexandre A Steiner
ABSTRACT
We tested the hypothesis that development of hypothermia instead of fever in endotoxic shock is consequential to hypoxia. Endotoxic shock was induced by bacterial lipopolysaccharide (LPS, 500 μg kg(-1) i.v.) in rats at an ambient temperature of 22 °C. A β3-adrenergic agonist known to activate metabolic heat production, CL316,243, was employed to evaluate whether thermogenic capacity could be impaired by the fall in oxygen delivery (ḊO2) during endotoxic shock. This possibility was rejected as CL316,243 (0.15 mg kg(-1) i.v.) evoked similar rises in oxygen consumption (V̇O2) in the presence and absence of endotoxic shock. Next, to investigate whether a less severe form of circulatory hypoxia could be triggering hypothermia, the circulating volume of LPS-injected rats was expanded using 6% hetastarch with the intention of improving tissue perfusion and alleviating hypoxia. This intervention attenuated not only the fall in arterial pressure induced by LPS, but also the associated falls in V̇O2 and body temperature. These effects, however, occurred independently of hypoxia, as they were not accompanied by any detectable changes in NAD(+)/NADH ratios. Further experimentation revealed that even the earliest drops in cardiac output and ḊO2 during endotoxic shock did not precede the reduction in V̇O2 that brings about hypothermia. In fact, ḊO2 and V̇O2 fell in such a synchrony that the ḊO2/V̇O2 ratio remained unaffected. Only when hypothermia was prevented by exposure to a warm environment (30 °C) did an imbalance in the ḊO2/V̇O2 ratio become evident, and such an imbalance was associated with reductions in the renal and hypothalamic NAD(+)/NADH ratios. In conclusion, hypometabolism and hypothermia in endotoxic shock are not consequential to hypoxia but serve as a pre-emptive strategy to avoid hypoxia in this model.
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Human Tumor Necrosis Factor α ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, natural, 97%, FG
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Bovine TNFα / Tumor Necrosis Factor alpha ELISA Kit, for serum, plasma and cell culture supernatants