  • Search Results
  • Polyethylene glycol-fused allografts produce rapid behavioral recovery after ablation of sciatic nerve segments.
Polyethylene glycol-fused allografts produce rapid behavioral recovery after ablation of sciatic nerve segments.
Journal of neuroscience research (2014-11-27)
D C Riley, G D Bittner, M Mikesh, N L Cardwell, A C Pollins, C L Ghergherehchi, S R Bhupanapadu Sunkesula, T N Ha, B T D Hall, A D Poon, M Pyarali, R B Boyer, A T Mazal, N Munoz, R C Trevino, T Schallert, W P Thayer
ABSTRACT
Restoration of neuronal functions by outgrowths regenerating at ∼1 mm/day from the proximal stumps of severed peripheral nerves takes many weeks or months, if it occurs at all, especially after ablation of nerve segments. Distal segments of severed axons typically degenerate in 1-3 days. This study shows that Wallerian degeneration can be prevented or retarded, and lost behavioral function can be restored, following ablation of 0.5-1-cm segments of rat sciatic nerves in host animals. This is achieved by using 0.8-1.1-cm microsutured donor allografts treated with bioengineered solutions varying in ionic and polyethylene glycol (PEG) concentrations (modified PEG-fusion procedure), being careful not to stretch any portion of donor or host sciatic nerves. The data show that PEG fusion permanently restores axonal continuity within minutes, as initially assessed by action potential conduction and intracellular diffusion of dye. Behavioral functions mediated by the sciatic nerve are largely restored within 2-4 weeks, as measured by the sciatic functional index. Increased restoration of sciatic behavioral functions after ablating 0.5-1-cm segments is associated with greater numbers of viable myelinated axons within and distal to PEG-fused allografts. Many such viable myelinated axons are almost certainly spared from Wallerian degeneration by PEG fusion. PEG fusion of donor allografts may produce a paradigm shift in the treatment of peripheral nerve injuries.
Sigma-Aldrich
Methylene blue, certified by the Biological Stain Commission
Sigma-Aldrich
Methylene Blue solution, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, for microscopy
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Supelco
Acetone, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)